Gene Transcripts

Gene Symbol
Nxf5
Entrez Gene ID
680025
Gene Name
nuclear RNA export factor 5
Gene Aliases
Chromosome Location
Chr. X: 106218058 - 106253226 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008773496.1 XP_008771718.1 10 976

View Full Product Details