Gene Transcripts

Gene Symbol
Uggt2
Entrez Gene ID
361091
Gene Name
UDP-glucose glycoprotein glucosyltransferase 2
Gene Aliases
RGD1563520
Chromosome Location
Chr. 15: 104235678 - 104405313 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006252469.2 XP_006252531.1 26, 27 3744

View Full Product Details