Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001013205.2 | NP_001013223.2 | 5, 6 | 486 | |
XM_006234930.1 | XP_006234992.1 | 5, 6 | 612 | ||
XM_006234931.2 | XP_006234993.1 | 5, 6 | 612 | ||
XM_006234932.2 | XP_006234994.1 | 5, 6 | 612 | ||
XM_008762198.1 | XP_008760420.1 | 5, 6 | 612 |