Gene Transcripts

Gene Symbol
Emilin3
Entrez Gene ID
362262
Gene Name
elastin microfibril interfacer 3
Gene Aliases
Chromosome Location
Chr. 3: 156899892 - 156906566 on Build Rnor_6.0
UniGene ID
Rn.141391
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109901.2 NP_001103371.1 2 346
XM_006235495.1 XP_006235557.1 2 1020

View Full Product Details