Gene Transcripts

Gene Symbol
Clec2g
Entrez Gene ID
362447
Gene Name
C-type lectin domain family 2, member G
Gene Aliases
CLEC2D11, Clr-b, Clr11, Clrb, RGD1563148
Chromosome Location
Chr. 4: 162934195 - 162942891 on Build Rnor_6.0
UniGene ID
Rn.202840
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001048075.1 NP_001041540.1 5 503
GenBank mRNA DQ168417.1 - 5 503
DQ168419.1 - 5 503
EF100690.1 - 5 503
EU128748.1 - 5 509

View Full Product Details