Gene Transcripts

Gene Symbol
Heatr5b
Entrez Gene ID
362683
Gene Name
HEAT repeat containing 5B
Gene Aliases
Chromosome Location
Chr. 6: 1330314 - 1410339 on Build Rnor_6.0
UniGene ID
Rn.6591
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191064.1 NP_001177993.1 8 1056

View Full Product Details