Gene Transcripts

Gene Symbol
Tigit
Entrez Gene ID
363784
Gene Name
T cell immunoreceptor with Ig and ITIM domains
Gene Aliases
RGD1563191
Chromosome Location
Chr. 11: 61970617 - 61987749 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006248323.2 XP_006248385.1 4 1103
XM_008768765.1 XP_008766987.1 4 1086

View Full Product Details