Gene Transcripts

Gene Symbol
Ido2
Entrez Gene ID
681319
Gene Name
indoleamine 2,3-dioxygenase 2
Gene Aliases
Indol1
Chromosome Location
Chr. 16: 72232852 - 72291227 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008771375.1 XP_008769597.1 10, 11 856

View Full Product Details