Gene Transcripts

Gene Symbol
Ero1lb
Entrez Gene ID
364755
Gene Name
ERO1-like beta (S. cerevisiae)
Gene Aliases
RGD1563548
Chromosome Location
Chr. 17: 90709394 - 90781848 on Build Rnor_6.0
UniGene ID
Rn.22313
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001191893.1 NP_001178822.1 11 761
XM_006254375.2 XP_006254437.1 11 951
XM_008771876.1 XP_008770098.1 11 727

View Full Product Details