Gene Transcripts

Gene Symbol
Olr34
Entrez Gene ID
404894
Gene Name
olfactory receptor 34
Gene Aliases
Chromosome Location
Chr. 1: 150310319 - 150311257 on Build Rnor_6.0
UniGene ID
Rn.141940
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000689.1 NP_001000689.1 1 442

View Full Product Details