Gene Transcripts

Gene Symbol
Olr96
Entrez Gene ID
405910
Gene Name
olfactory receptor 96
Gene Aliases
Chromosome Location
Chr. 1: 168489077 - 168490033 on Build Rnor_6.0
UniGene ID
Rn.141891
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001001025.1 NP_001001025.1 1 292

View Full Product Details