Gene Transcripts

Gene Symbol
Eltd1
Entrez Gene ID
64124
Gene Name
EGF, latrophilin and seven transmembrane domain containing 1
Gene Aliases
Etl
Chromosome Location
Chr. 2: 256609787 - 256724642 on Build Rnor_6.0
UniGene ID
Rn.8731
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_022294.1 NP_071630.1 10 1521
GenBank mRNA AF192401.1 - 10 1521
AF192402.1 - 10 1521

View Full Product Details