Gene Transcripts

Gene Symbol
Ppap2b
Entrez Gene ID
192270
Gene Name
phosphatidic acid phosphatase type 2B
Gene Aliases
Dri42
Chromosome Location
Chr. 5: 124690214 - 124765499 on Build Rnor_6.0
UniGene ID
Rn.12038
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138905.2 NP_620260.2 5 1138
GenBank mRNA BC072544.1 - 5 1138
Y07783.1 - 5 1130

View Full Product Details