Gene Transcripts

Gene Symbol
Mc1r
Entrez Gene ID
102552838
Gene Name
melanocortin 1 receptor
Gene Aliases
Chromosome Location
Chr. 19: 56215421 - 56219987 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006255795.2 XP_006255857.1 1 1974

View Full Product Details