Gene Transcripts

Gene Symbol
Elane
Entrez Gene ID
299606
Gene Name
elastase, neutrophil expressed
Gene Aliases
Ela2
Chromosome Location
Chr. 7: 12638320 - 12640168 on Build Rnor_6.0
UniGene ID
Rn.90573
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106767.1 NP_001100237.1 4 527

View Full Product Details