Gene Transcripts

Gene Symbol
Pabpc1l2a
Entrez Gene ID
302405
Gene Name
poly(A) binding protein, cytoplasmic 1-like 2A
Gene Aliases
Pabpc1l2b, RGD1566367, Rbm32b
Chromosome Location
Chr. X: 73894616 - 73895719 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006257150.2 XP_006257212.1 1 463

View Full Product Details