Gene Transcripts

Gene Symbol
Fmo6
Entrez Gene ID
304922
Gene Name
flavin containing monooxygenase 6
Gene Aliases
RGD1559491
Chromosome Location
Chr. 13: 80791355 - 80819672 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006250206.2 XP_006250268.1 2 164

View Full Product Details