Gene Transcripts

Gene Symbol
Olr29
Entrez Gene ID
404896
Gene Name
olfactory receptor 29
Gene Aliases
Chromosome Location
Chr. 1: 150209396 - 150210403 on Build Rnor_6.0
UniGene ID
Rn.141943
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000691.1 NP_001000691.1 1 49

View Full Product Details