Gene Transcripts

Gene Symbol
Mk1
Entrez Gene ID
171436
Gene Name
Mk1 protein
Gene Aliases
Chromosome Location
Chr. 16: 20531720 - 20534209 on Build Rnor_6.0
UniGene ID
Rn.201717
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_134399.2 NP_599226.1 1 2028

View Full Product Details