Gene Transcripts

Gene Symbol
Slc22a25
Entrez Gene ID
192273
Gene Name
solute carrier family 22, member 25
Gene Aliases
Slc22a9, Ust1, Ust1r
Chromosome Location
Chr. 1: 224596886 - 224698512 on Build Rnor_6.0
UniGene ID
Rn.31066
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_138908.2 NP_620263.1 7 1303
GenBank mRNA Y09945.1 - 7 1305

View Full Product Details