Gene Transcripts

Gene Symbol
Rbfa
Entrez Gene ID
307235
Gene Name
ribosome binding factor A
Gene Aliases
RGD1311910
Chromosome Location
Chr. 18: 76704223 - 76714384 on Build Rnor_6.0
UniGene ID
Rn.105128
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107373.1 NP_001100843.1 7 737
XM_006254951.2 XP_006255013.1 6 842

View Full Product Details