Gene Transcripts

Gene Symbol
Taco1
Entrez Gene ID
360645
Gene Name
translational activator of mitochondrially encoded cytochrome c oxidase I
Gene Aliases
Ccdc44, RGD1306784
Chromosome Location
Chr. 10: 94260148 - 94268054 on Build Rnor_6.0
UniGene ID
Rn.137785
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108302.1 NP_001101772.1 1, 2 486
GenBank mRNA BC166900.1 - 1, 2 437

View Full Product Details