Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001134409.1 | NP_001127881.1 | 7, 8 | 1147 | |
XM_008761077.1 | XP_008759299.1 | 8, 9 | 1320 | ||
GenBank mRNA | X99773.1 | - | 7, 8 | 1162 | |
Z30585.1 | - | 5, 6 | 515 |