Gene Transcripts

Gene Symbol
Kcnq5
Entrez Gene ID
259273
Gene Name
potassium channel, voltage-gated KQT-like subfamily Q, member 5
Gene Aliases
Kcnq5l
Chromosome Location
Chr. 9: 27565869 - 28141114 on Build Rnor_6.0
UniGene ID
Rn.32232
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001134643.2 NP_001128115.2 4 801
GenBank mRNA FQ084615.1 - 4 442

View Full Product Details