Gene Transcripts

Gene Symbol
Vom2r31
Entrez Gene ID
286985
Gene Name
vomeronasal 2 receptor, 31
Gene Aliases
Chromosome Location
Chr. 1: 74975364 - 75001212 on Build Rnor_6.0
UniGene ID
Rn.10812
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173319.1 NP_775441.1 3 1172
GenBank mRNA AF016184.1 - 3 1172

View Full Product Details