Gene Transcripts

Gene Symbol
Ugt2b7
Entrez Gene ID
286989
Gene Name
UDP glucuronosyltransferase 2 family, polypeptide B7
Gene Aliases
UDPGT 2B7, Ugt2b8
Chromosome Location
Chr. 14: 22597103 - 22619968 on Build Rnor_6.0
UniGene ID
Rn.11131
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_173323.1 NP_775445.1 1 468
GenBank mRNA U27518.1 - 1 468

View Full Product Details