Gene Transcripts

Gene Symbol
Sectm1a
Entrez Gene ID
287885
Gene Name
secreted and transmembrane 1A
Gene Aliases
Sectm1
Chromosome Location
Chr. 10: 110263782 - 110274795 on Build Rnor_6.0
UniGene ID
Rn.145435
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001013043.1 NP_001013061.1 3 500
XM_006247885.2 XP_006247947.1 3 527
GenBank mRNA BC078937.1 - 3 500
FQ225276.1 - 3 493

View Full Product Details