Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001134990.1 | NP_001128462.1 | 2 | 272 | |
XM_006252059.2 | XP_006252121.1 | 3 | 1125 | ||
XM_006252061.2 | XP_006252123.1 | 2 | 953 | ||
GenBank mRNA | BC168660.1 | - | 2 | 272 | |
FQ230419.1 | - | 3 | 322 |