Gene Transcripts

Gene Symbol
Pcdhb3
Entrez Gene ID
291656
Gene Name
protocadherin beta 3
Gene Aliases
Chromosome Location
Chr. 18: 30381322 - 30383876 on Build Rnor_6.0
UniGene ID
Rn.214506
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014783.2 NP_001014783.1 1 1153

View Full Product Details