Gene Transcripts

Gene Symbol
Psx1
Entrez Gene ID
296535
Gene Name
placenta specific homeobox 1
Gene Aliases
Chromosome Location
Chr. 3: 573921 - 574592 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_231064.2 XP_231064.2 1 552

View Full Product Details