Gene Transcripts

Gene Symbol
Cenpa
Entrez Gene ID
298850
Gene Name
centromere protein A
Gene Aliases
RGD1563607
Chromosome Location
Chr. 6: 27072262 - 27080861 on Build Rnor_6.0
UniGene ID
Rn.16165
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106711.1 NP_001100181.1 4 376
XM_006239798.2 XP_006239860.1 4 499
GenBank mRNA BC166997.1 - 4 417

View Full Product Details