Gene Transcripts

Gene Symbol
Trim43a
Entrez Gene ID
300884
Gene Name
tripartite motif-containing 43A
Gene Aliases
RGD1561074
Chromosome Location
Chr. 8: 96389344 - 96401733 on Build Rnor_6.0
UniGene ID
Rn.198201
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001142944.1 NP_001136416.1 3, 4 740
XM_006243512.2 XP_006243574.1 4, 5 980

View Full Product Details