Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001025670.1 | NP_001020841.1 | 11 | 1463 | |
XM_008770019.1 | XP_008768241.1 | 12 | 1460 | ||
XM_008770020.1 | XP_008768242.1 | 12 | 1889 | ||
XM_008770021.1 | XP_008768243.1 | 12 | 1588 | ||
GenBank mRNA | BC097362.1 | - | 11 | 1463 |