Gene Transcripts

Gene Symbol
Nip30
Entrez Gene ID
307652
Gene Name
NEFA-interacting nuclear protein NIP30
Gene Aliases
RGD1307433
Chromosome Location
Chr. 19: 10827057 - 10856747 on Build Rnor_6.0
UniGene ID
Rn.6506
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014014.1 NP_001014036.1 4 464
XM_006255110.2 XP_006255172.1 4 774
XM_006255111.2 XP_006255173.1 4 541
XM_006255114.2 XP_006255176.1 5 1006
GenBank mRNA BC079145.1 - 4 464

View Full Product Details