Gene Transcripts

Gene Symbol
Nkd2
Entrez Gene ID
308068
Gene Name
naked cuticle homolog 2 (Drosophila)
Gene Aliases
Chromosome Location
Chr. 1: 32054412 - 32085416 on Build Rnor_6.0
UniGene ID
Rn.198099
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107454.1 NP_001100924.1 3, 4 343
XM_008758692.1 XP_008756914.1 6, 7 2077
XM_008758693.1 XP_008756915.1 3, 4 858

View Full Product Details