Gene Transcripts

Gene Symbol
Fam46c
Entrez Gene ID
310721
Gene Name
family with sequence similarity 46, member C
Gene Aliases
Chromosome Location
Chr. 2: 202796500 - 202816562 on Build Rnor_6.0
UniGene ID
Rn.162620
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001014041.1 NP_001014063.1 2 483
XM_006233038.2 XP_006233100.1 2 301
XM_006233039.2 XP_006233101.1 2 405
XM_008761361.1 XP_008759583.1 2 273
GenBank mRNA BC083570.1 - 2 483

View Full Product Details