Gene Transcripts

Gene Symbol
Zyg11a
Entrez Gene ID
313482
Gene Name
zyg-11 family member A, cell cycle regulator
Gene Aliases
RGD1309582
Chromosome Location
Chr. 5: 127790496 - 127828911 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_233348.8 XP_233348.6 9 1658

View Full Product Details