Gene Transcripts

Gene Symbol
Hs6st3
Entrez Gene ID
364476
Gene Name
heparan sulfate 6-O-sulfotransferase 3
Gene Aliases
RGD1560050
Chromosome Location
Chr. 15: 104448479 - 104452907 on Build Rnor_6.0
UniGene ID
Rn.234606
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001271404.1 NP_001258333.1 1189

View Full Product Details