Gene Transcripts

Gene Symbol
Syne2
Entrez Gene ID
366669
Gene Name
spectrin repeat containing, nuclear envelope 2
Gene Aliases
RGD1305248
Chromosome Location
Chr. 6: 98884269 - 99153551 on Build Rnor_6.0
UniGene ID
Rn.50877
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008764911.1 XP_008763133.1 107 19951
GenBank mRNA BC098809.1 - 5 648

View Full Product Details