Gene Transcripts

Gene Symbol
Klk5l
Entrez Gene ID
408211
Gene Name
kallikrein 5-like
Gene Aliases
KAL, KALA, Klk1, Klk5, RATKALA
Chromosome Location
Chr. 1: 100055975 - 100058310 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759478.1 XP_008757700.1 3 515

View Full Product Details