Gene Transcripts

Gene Symbol
Vom1r58
Entrez Gene ID
494226
Gene Name
vomeronasal 1 receptor 58
Gene Aliases
V1rj6
Chromosome Location
Chr. 1: 75131348 - 75132167 on Build Rnor_6.0
UniGene ID
Rn.129410
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009507.1 NP_001009507.1 1 83
GenBank mRNA AY510331.1 - 1 83

View Full Product Details