Gene Transcripts

Gene Symbol
Vom1r21
Entrez Gene ID
494229
Gene Name
vomeronasal 1 receptor 21
Gene Aliases
V1rf3
Chromosome Location
Chr. 19: 28377249 - 28378118 on Build Rnor_6.0
UniGene ID
Rn.129448
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009510.1 NP_001009510.1 1 23
GenBank mRNA AY510339.1 - 1 23

View Full Product Details