Gene Transcripts

Gene Symbol
Vom1r72
Entrez Gene ID
494241
Gene Name
vomeronasal 1 receptor 72
Gene Aliases
V1rc39
Chromosome Location
Chr. 4: 87699732 - 87700631 on Build Rnor_6.0
UniGene ID
Rn.129668
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009521.1 NP_001009521.1 1 417
GenBank mRNA AY510329.1 - 1 417

View Full Product Details