Gene Transcripts

Gene Symbol
Vom1r85
Entrez Gene ID
494250
Gene Name
vomeronasal 1 receptor 85
Gene Aliases
V1rc44
Chromosome Location
Chr. 4: 88232009 - 88232917 on Build Rnor_6.0
UniGene ID
Rn.129655
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001009530.1 NP_001009530.1 1 596
GenBank mRNA AY510325.1 - 1 596

View Full Product Details