Gene Transcripts

Gene Symbol
Vom1r47
Entrez Gene ID
494266
Gene Name
vomeronasal 1 receptor 47
Gene Aliases
V1rg14, V1rg9
Chromosome Location
Chr. 1: 67025240 - 67026154 on Build Rnor_6.0
UniGene ID
Rn.129420
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008916.1 NP_001008916.1 1 402
GenBank mRNA AY510316.1 - 1 402
AY510317.1 - 1 402

View Full Product Details