Gene Transcripts

Gene Symbol
Vom1r32
Entrez Gene ID
494284
Gene Name
vomeronasal 1 receptor 32
Gene Aliases
V1rd24
Chromosome Location
Chr. 1: 71844368 - 71845285 on Build Rnor_6.0
UniGene ID
Rn.129432
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008937.1 NP_001008937.1 1 795
GenBank mRNA AY510337.1 - 1 795

View Full Product Details