Gene Transcripts

Gene Symbol
Xkr9
Entrez Gene ID
497099
Gene Name
XK, Kell blood group complex subunit-related family, member 9
Gene Aliases
XRG9
Chromosome Location
Chr. 5: 4946332 - 4975436 on Build Rnor_6.0
UniGene ID
Rn.161960
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001012229.1 NP_001012229.1 4 1125
XM_006237733.2 XP_006237795.1 4 1208
GenBank mRNA AY534263.1 - 4 1125

View Full Product Details