Gene Transcripts

Gene Symbol
Vgll3
Entrez Gene ID
498038
Gene Name
vestigial-like family member 3
Gene Aliases
RGD1560030
Chromosome Location
Chr. 11: 3119505 - 3168888 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006247980.2 XP_006248042.1 4 1374
XM_006247981.2 XP_006248043.1 4 1372
XM_006247982.2 XP_006248044.1 4 1204

View Full Product Details