Gene Transcripts

Gene Symbol
Tmprss11f
Entrez Gene ID
498345
Gene Name
transmembrane protease, serine 11f
Gene Aliases
RGD1560675
Chromosome Location
Chr. 14: 23982415 - 24055137 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008770104.1 XP_008768326.1 5 452

View Full Product Details