Gene Transcripts

Gene Symbol
Lpar2
Entrez Gene ID
498609
Gene Name
lysophosphatidic acid receptor 2
Gene Aliases
Edg4, Pbx4, RGD1561336
Chromosome Location
Chr. 16: 21343313 - 21348913 on Build Rnor_6.0
UniGene ID
Rn.14855
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109109.1 NP_001102579.1 2 302

View Full Product Details